Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YAVQIHILKA DKAGDWWKFT VNIISVYKQG TSRIRRGDQS LWIRSRDIAC KCPKIKPLKK YLLLGNAEDS PDQSGIVADK SSLVIQWRDT WARRLRKFQQ REKKGKCKKA
Netrin-1 is a protein target that plays a crucial role in various biological processes, particularly in the development and guidance of neuronal cells. It belongs to the netrin family of axon guidance molecules and is involved in mediating axon guidance, cell migration, and tissue morphogenesis during embryonic development. Netrin-1 functions by binding to specific receptors on the cell surface and providing directional cues for cellular movement and connectivity. Dysregulation of Netrin-1 has been implicated in various diseases, including cancer, neurological disorders, and cardiovascular conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epididymis tissue protein Li 131P; HGNC:8029; netrin 1, mouse, homolog of; Netrin-1; Netrin1
Gene Aliases: NTN1; NTN1L
UniProt ID: (Human) O95631
Entrez Gene ID: (Human) 9423
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support