Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: PIIGVVENMS GFICPKCKKE SQIFPPTTGG AELMCQDLEV PLLGRVPLDP LIGKNCDKGQ SFFIDAPDSP ATLAYRSIIQ RIQEFCNLHQ SKEENLISS
Component of the cytosolic iron-sulfur (Fe/S) protein assembly (CIA) machinery (PubMed:18573874). Required for maturation of extramitochondrial Fe-S proteins (PubMed:18573874). The NUBP1-NUBP2 heterotetramer forms a Fe-S scaffold complex, mediating the de novo assembly of an Fe-S cluster and its transfer to target apoproteins (PubMed:18573874). Implicated in the regulation of centrosome duplication. Negatively regulates cilium formation and structure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytosolic Fe-S cluster assembly factor NUBP1; NBP 1; NUBP1; nucleotide binding protein (e.coli MinD like); nucleotide binding protein 1 (E.coli MinD like); nucleotide binding protein 1 (MinD homolog, E. coli); Nucleotide-binding protein 1
Gene Aliases: NBP; NBP1; NBP35; NUBP1
UniProt ID: (Human) P53384
Entrez Gene ID: (Human) 4682
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support