Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: KATEKKEFLE PDSWETLDQQ QFFTEEELKE YENIIALQEN ELKKKADELQ KQKEELQRQH DQLEAQKLEY HQVIQQMEQK KLQQGIPPSG PAGELKFEPH I
NUCB2 was initially identified as calcium-binding protein EF hand motif-containing protein that could bind with Necdin, a growth suppressor expressed primarily in postmitotic neurons. NUC2 is a secreted protein that is cleaved into three major peptide products, Nesfatin-1, Nesfatin-2, and Nesfatin-3, of which Nesfatin-1 has been found to cause the suppression of food intake in a leptin-independent manner. Other studies have suggested that NUCB2/Nesfatin-1 may also play roles in energy homeostasis and closely related neuroendocrine functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-binding protein NEFA; Epididymis secretory protein Li 109; Gastric cancer antigen Zg4; nesfatin 1; nesfatin-1; Nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobinding 2; prepronefastin; Prepronesfatin
Gene Aliases: AI607786; Calnuc; HEL-S-109; NEFA; Nesfatin-1; NUCB2; p54
UniProt ID: (Human) P80303, (Mouse) P81117, (Rat) Q9JI85
Entrez Gene ID: (Human) 4925, (Mouse) 53322, (Rat) 59295
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support