Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRPVRNRKV VDYSQFQESD DADEDYGRDS GPPTKKIRSS PREAKNKRRS GKNSQEDSED SEDKDVKTKK DDSHSAEDSE DEKEDHKNVR QQRQAASKAA SKQREMLMED VGSEEEQEEE DEAPFQEKDS GSDEDFLMED DDDSDYGSSK KKNKKMVKKS KPERKEKKMP KPRLKATVTP SPVKGKGKVG RPTASKASKE KTPSPKEEDE EPESPPEKKT STSPPPEKSG DEGSEDEAPS GED
This gene encodes a nuclear protein that is highly conserved in vertebrates. The conserved regions of the protein contain several consensus phosphorylation sites for casein kinase II and cyclin-dependent kinases, two putative nuclear localization signals, and a basic DNA-binding domain. It is phosphorylated in vivo by Cdk1 during mitosis of the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nuclear casein kinase and cyclin-dependent kinase substrate 1; Nuclear ubiquitous casein and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate; P1; potential LAG1 interactor
Gene Aliases: JC7; NUCKS; NUCKS1
UniProt ID: (Human) Q9H1E3
Entrez Gene ID: (Human) 64710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support