Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MSSVKRSLKQ EIVTQFHCSA AEGDIAKLTG ILSHSPSLLN ETSENGWTAL MYAARNGHPE IVQFLLEKGC DRSIVNKSRQ TALDIAVFWG YKHIANLLAT AKGGKKPWFL TNEVEECENY FSKTLLDRKS EKRNNSDWLL AKESHPATVF ILFSDLNPLV TLGGNKESFQ QPEVRLCQLN YTDIKDYLAQ PEKITLIFLG VELEIKDKLL NYAGEVPREE EDGLVAWFAL GIDPIAAEEF KQRHENCYFL HPPMPALLQL KEKEAGVVAQ ARSVLAWHSR YKFCPTCGNA TKIEEGGYKR LCLKEDCPSL NGVHNTSYPR VDPVVIMQVI HPDGTKCLLG RQKRFPPGMF TCLAGFIEPG ETIEDAVRRE VEEESGVKVG HVQYVACQPW PMPSSLMIGC LALAVSTEIK VDKNEIEDAR WFTREQVLDV LTKGKQQAFF VPPSRAIAHQ LIKHWIRINP NL
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DeNADding enzyme NUDT12; DKFZP761I172; NAD-capped RNA hydrolase NUDT12; NADH pyrophosphatase NUDT12; nucleoside diphosphate linked moiety X-type motif 12; Nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; Nudix motif 12
Gene Aliases: NUDT12
UniProt ID: (Human) Q9BQG2
Entrez Gene ID: (Human) 83594
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support