Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • NUDT2 Antibodies

            Proteintech

            NUDT2 Polyclonal Antibody

            View all (10) NUDT2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # 10484-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Product not available in your region
            150 µL
             
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite NUDT2 Polyclonal Antibody

            • Antibody Testing Data (11)
            Application
            NUDT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 11

            NUDT2 Antibody (10484-1-AP) in IHC (P)

            Immunohistochemistry of paraffin-embedded human breast cancer tissue slide using 10484-1-AP (NUDT2 antibody) at dilution of 1:600 (under 40x lens) heat mediated antigen retrieved with Tris-EDTA buffer (pH 9).
            View Product

            NUDT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            NUDT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            NUDT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            NUDT2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            NUDT2 Antibody in Western Blot (WB)
            NUDT2 Antibody in Western Blot (WB)
            NUDT2 Antibody in Western Blot (WB)
            NUDT2 Antibody in Western Blot (WB)
            NUDT2 Antibody in Western Blot (WB)
            NUDT2 Antibody in Western Blot (WB)
            NUDT2 Antibody in Immunoprecipitation (IP)

            Product Details

            10484-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:3000-1:8000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:200-1:600
            -

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            NUDT2 Fusion Protein Ag0743 (1-149 aa encoded by BC004926)

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MALRACGLI IFRRCLIPKV DNNAIEFLLL QASDGIHHWT PPKGHVEPGE DDLETALRET QEEAGIEAGQ LTIIEGFKRE LNYVARNKPK TVIYWLAEVK DYDVEIRLSH EHQAYRWLGL EEACQLAQFK EMKAALQEGH QFLCSIEA (1-149 aa encoded by BC004926)

            Target Information

            This gene encodes a member of the MutT family of nucleotide pyrophosphatases, a subset of the larger NUDIX hydrolase family. The gene product possesses a modification of the MutT sequence motif found in certain nucleotide pyrophosphatases. The enzyme asymmetrically hydrolyzes Ap4A to yield AMP and ATP and is responsible for maintaining the intracellular level of the dinucleotide Ap4A, the function of which has yet to be established. This gene may be a candidate tumor suppressor gene. Alternative splicing has been observed at this locus and three transcript variants, all encoding the same protein, have been identified.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Ap4A hydrolase; Ap4A hydrolase 1; Ap4Aase; bis(5'-nucleosyl)-tetraphosphatase; bis(5'-nucleosyl)-tetraphosphatase (asymmetrical); Bis(5'-nucleosyl)-tetraphosphatase [asymmetrical]; Diadenosine 5',5'''-P1,P4-tetraphosphate asymmetrical hydrolase; diadenosine 5',5''-P1,P4-tetraphosphate pyrophosphohydrolase; diadenosine tetraphosphatase; MGC10404; Nucleoside diphosphate-linked moiety X motif 2; nudix (nucleoside diphosphate linked moiety X)-type motif 2; Nudix motif 2

            View more View less

            Gene Aliases: 2310051L06Rik; AA939917; APAH1; NUDT2

            View more View less

            UniProt ID: (Human) P50583, (Mouse) P56380, (Rat) Q6PEC0

            View more View less

            Entrez Gene ID: (Human) 318, (Mouse) 66401, (Rat) 297998

            View more View less

            Function(s)

            bis(5'-nucleosyl)-tetraphosphatase (asymmetrical) activity protein binding GTP binding bis(5'-nucleosyl)-tetraphosphatase (symmetrical) activity nucleotide binding bis(5'-nucleosyl)-tetraphosphatase activity hydrolase activity hydrolase metabolite interconversion enzyme

            Process(es)

            response to reactive oxygen species nucleobase-containing compound metabolic process apoptotic process metabolic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            • Fair Trade
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            korea | 대한민국 flag icon
            Korea | 대한민국

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Thermo Fisher Scientific Korea Ltd.
            Representative : Soojin Seok
            Company Registration No. : 117-81-46910

             

            Thermo Fisher Scientific Solutions LLC
            Representative : Soojin Seok
            Company Registration No. : 114-86-04783

             

            Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-7765t:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0