Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNKLRQSLR RRKPAYVPEA SRPHQWQADE DAVRKGTCSF PVRYLGHVEV EESRGMHVCE DAVKKLKAMG RKSVKSVLWV SADGLRVVDD KTKDLLVDQT IEKVSFCAPD RNLDKAFSYI CRDGTTRRWI CHCFLALKDS GERLSHAVGC AFAACLERKQ RREKECGVTA AFDASRTSFA REGSFRLSGG GRPAEREAPD KKKAEAAAAP TVAPGPAQPG HVSPTPATTS PGEKGEAGTP VAAGTTAAAI PRRHAPLEQL VRQGSFRGFP ALSQKNSPFK R (1-280 aa encoded by BC001794)
Plays a role in the process of neurogenesis. Required throughout embryonic neurogenesis to maintain neural progenitor cells, also called radial glial cells (RGCs), by allowing their daughter cells to choose progenitor over neuronal cell fate. Not required for the proliferation of neural progenitor cells before the onset of embryonic neurogenesis. Also required postnatally in the subventricular zone (SVZ) neurogenesis by regulating SVZ neuroblasts survival and ependymal wall integrity. Negative regulator of NF-kappa-B signaling pathway. The inhibition of NF-kappa-B activation is mediated at least in part, by preventing MAP3K7IP2 to interact with polyubiquitin chains of TRAF6 and RIPK1 and by stimulating the 'Lys-48'-linked polyubiquitination and degradation of TRAF6 in cortical neurons. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: numb homolog (Drosophila)-like; numb homolog-like; Numb-like protein; Numb-R; Numb-related protein
Gene Aliases: CAG3A; CTG3a; NBL; NUMB-R; NUMBL; NUMBLIKE; NUMBR; TNRC23
UniProt ID: (Human) Q9Y6R0, (Mouse) O08919
Entrez Gene ID: (Human) 9253, (Mouse) 18223
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support