Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: A375, LO2, HeLa, NCI-H460. The target is usually found in the following locations: Chromosome, Nucleus, centromere, kinetochore, nuclear pore complex.
Immunogen sequence: NILKDMLQAA SHYRQNRNSL YRREESLEKE PEYVPWTATS GPGGIRTVII RQHEIVLKVA YPQADSNLRN IVTEQLVALI DCFLDGYVSQ LKSVDKSSNR ERYDNLEMEY LQKRSDLLSP LLSLGQYLWA ASLAEKYCDF DILVQMCEQT DNQSRLQRYM TQFADQNFSD FLFRWYLEKG KRGKLLSQPI SQHGQLANFL Q
Plays a role in T-cell signaling pathway. Isoform 2 may play a role in T-helper 2 (Th2) signaling pathway and seems to represent the first proximal signaling protein that links T-cell receptor-mediated signal to the activation of c-Maf Th2 specific factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 133 kDa nucleoporin; Nuclear pore complex protein Nup133; nucleoporin 133kD; nucleoporin 133kDa; Nucleoporin Nup133
Gene Aliases: hNUP133; NUP133
UniProt ID: (Human) Q8WUM0
Entrez Gene ID: (Human) 55746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support