Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LIQVDLADK LLQVASPFLE PHLVRMAKVD QNRVRYMDLL WRYYEKNRSF SNAARVLSRL ADMHSTEISL QQRLEYIARA ILSAKSSTAI SSIAADGEFL HELEEKMEVA RIQLQIQETL QRQYSHHSSV QDAVSQLDSE LMDITKLYGE FADPFKLAEC KLAIIHCAGY SDPILVQTLW QDIIEKELSD SVTLSSSDRM HALSLKIVLL GKIYAGTPRF FPLDFIVQFL EQQVCTLNWD VGFVIQTMNE IGVPLPRLLE VYDQLFKSRD PFWNRMKKPL HLLDCIHVLL IRYVENPSQV LNCERRRFTN LCLDAVCGYL VELQSMSSSV AVQAITGNFK SLQAKLERLH (1043-1391 aa encoded by BC039257)
Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 155 kDa nucleoporin; Nuclear pore complex protein Nup155; nucleoporin 155kD; nucleoporin 155kDa; Nucleoporin Nup155; P140
Gene Aliases: ATFB15; KIAA0791; N155; NUP155
UniProt ID: (Human) O75694, (Rat) P37199
Entrez Gene ID: (Human) 9631, (Rat) 117021
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support