Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is A549 cell lysate.
PA5-20565 can be used with blocking peptide PEP-0685.
The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-160 is part of the hetero-oligomeric complex that also contains NUP107, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP160 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP160 may be an important drug target in HIV treatment. Multiple isoforms of NUP160 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 160 kDa nucleoporin; Gene trap locus 1-13 protein; gene trap locus-13; GTL-13; Nuclear pore complex protein Nup160; nucleoporin 160kD; nucleoporin 160kDa; Nucleoporin Nup160
Gene Aliases: 160kDa; 2810011M03Rik; AA414952; AU020188; Gtl-13; Gtl1-13; Gtl13; KIAA0197; mKIAA0197; NUP120; NUP160
UniProt ID: (Human) Q12769, (Mouse) Q9Z0W3
Entrez Gene ID: (Human) 23279, (Mouse) 59015, (Rat) 311182
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support