Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MSGFNFGGTG APTGGFTFGT AKTATTTPAT GFSFSTSGTG GFNFGAPFQP ATSTPSTGLF SLATQTPATQ TTGFTFGTAT LASGGTGFSL GIGASKLNLS NTAATPAMAN PSGFGLGSSN LTNAISSTVT SSQGTAPTGF VFGPSTTSVA PATTSGGFSF TGGSTAQPSG FNIGSAGNSA QPTAPATLPF TPATPAATTA GATQPAAPTP TATITSTGPS LFASIATAPT SSATTGLSLC TPVTTAGAPT AGTQGFSLKA PGAASGTSTT TSTAATATAT TTSSSSTTGF ALNLKPLAPA GIPSNTAAAV TAPPGPGAAA GAAASSAMTY AQLESLINKW SLELEDQERH FLQQATQVNA WDRTLIENGE KITSLHREVE KVKLDQKRLD QELDFILSQQ KELEDLLSPL EELVKEQSGT IYLQHADEER EKTYKLAENI DAQLKRMAQD LKDIIEHLNT SGAPADTSDP LQQICKILNA HMDSLQWIDQ NSALLQRKVE EVTKVCEGRR KEQERSFRIT FD
SDS3 is a subunit of the histone deacetylase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 62 kDa nucleoporin; DKFZp547L134; FLJ20822; FLJ43869; MGC841; Nuclear pore glycoprotein p62; nucleoporin 62kD; nucleoporin 62kDa; Nucleoporin Nup62; nucleoporin p62
Gene Aliases: IBSN; NUP62; p62; SNDI
UniProt ID: (Human) P37198
Entrez Gene ID: (Human) 23636
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support