Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IGSAGNSAQ PTAPATLPFT PATPAATTAG ATQPAAPTPT ATITSTGPSL FASIATAPTS SATTGLSLCT PVTTAGAPTA GTQGFSLKAP GAASGTSTTT STAATATATT TSSSSTTGFA LNLKPLAPAG IPSNTAAAVT APPGPGAAAG AAASSAMTYA QLESLINKWS LELEDQERHF LQQATQVNAW DRTLIENGEK ITSLHREVEK VKLDQKRLDQ ELDFILSQQK ELEDLLSPLE ELVKEQSGTI YLQHADEERE KTYKLAENID AQLKRMAQDL KDIIEHLNTS GAPADTSDPL QQICKILNAH MDSLQWIDQN SALLQRKVEE VTKVCEGRRK EQERSFRITF D (173-522 aa encoded by BC014842)
SDS3 is a subunit of the histone deacetylase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 62 kDa nucleoporin; DKFZp547L134; FLJ20822; FLJ43869; MGC841; nuclear pore complex 1; nuclear pore glycoprotein 62; Nuclear pore glycoprotein p62; nucleoporin 62kD; nucleoporin 62kDa; Nucleoporin Nup62; nucleoporin p62
Gene Aliases: AA589433; AI426861; AI790512; AU045898; D7Ertd649e; IBSN; Np62; NUP62; Nupc1; p62; SNDI
UniProt ID: (Human) P37198, (Rat) P17955, (Mouse) Q63850
Entrez Gene ID: (Human) 23636, (Rat) 65274, (Mouse) 18226
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support