Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: IAERYRAQGI SANKFVDSTF YLLLDLITFF DEYHSGHIDR AFDIIERLKL VPLNQESVEE RVAAFRNFSD EIRHNLSEVL LATMNILFTQ FKRLKGTSPS SSSRPQRVIE DRDSQLRSQA RTLITFAGMI PYRTSGDTNA RLVQMEVLMN; Positive Samples: U-87MG, A431, LO2, HeLa, Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Rat testis; Cellular Location: Nucleus, Nucleus membrane, Peripheral membrane protein, nuclear pore complex
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. This gene encodes a nucleoporin protein that localizes both to the basket of the pore and to the nuclear entry of the central gated channel of the pore. The encoded protein is a target of caspase cysteine proteases that play a central role in programmed cell death by apoptosis. Alternative splicing results in multiple transcript variants encoding different isoforms.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 93 kDa nucleoporin; CBP-interacting protein 4; Nuclear pore complex protein Nup93; nucleoporin 93kDa; Nucleoporin Nup93
Gene Aliases: 2410008G02Rik; Cip4; KIAA0095; NIC96; NPHS12; NUP93
UniProt ID: (Human) Q8N1F7, (Mouse) Q8BJ71, (Rat) Q66HC5
Entrez Gene ID: (Human) 9688, (Mouse) 71805, (Rat) 291874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support