Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: SKPVDENHQQ DGDEDSLVSH FYTNPIAKPI PQTPESAGNK HSNSNSVDDT IVALNMRAAL RNGLEGSSEE TSFHDESLQD DREEIENNSY HMHPAGIILT KVGYYTIPSM DDLAKITNEK GECIVSDFTI GRKGYGSIYF EGDVNLTNLN LDDIVHIRRK EVVVYLDDNQ KPPVGEGLNR KAEVTLDGVW PTDKTSRCLI KSPDRLADIN YEGRLEAVSR KQGAQFKEYR PETGSWVFKV SHFSKYGLQD SDEEEEEHPS KTSTKKLKTA PLPPASQTTP LQMALNGKPA PPPQVEKKGQ; Positive Samples: A-549, LO2, HeLa, Jurkat, HL-60; Cellular Location: Nucleoplasmic side, Nucleus, Nucleus membrane, Peripheral membrane protein, nuclear pore complex
Signal-mediated nuclear import and export proceed through the nuclear pore complex (NPC), which is comprised of approximately 50 unique proteins collectively known as nucleoporins. The 98 kDa nucleoporin is generated through a biogenesis pathway that involves synthesis and proteolytic cleavage of a 186 kDa precursor protein. This cleavage results in the 98 kDa nucleoporin as well as a 96 kDa nucleoporin, both of which are localized to the nucleoplasmic side of the NPC. Rat studies show that the 98 kDa nucleoporin functions as one of several docking site nucleoporins of transport substrates. The human gene has been shown to fuse to several genes following chromosome translocations in acute myelogenous leukemia (AML) and T-cell acute lymphocytic leukemia (T-ALL). This gene is one of several genes located in the imprinted gene domain of 11p15. 5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in several transcript variants; however, not all variants have been fully described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GLFG-repeat containing nucleoporin; Nuclear pore complex protein Nup98-Nup96; nucleoporin 98kD; nucleoporin 98kDa; Nup98-Nup96; NUP98/PHF23 fusion 2 protein; OTTHUMP00000013954
Gene Aliases: 4732457F17; ADAR2; ADIR2; AI849286; NUP196; NUP96; NUP98
UniProt ID: (Human) P52948, (Mouse) Q6PFD9
Entrez Gene ID: (Human) 4928, (Mouse) 269966
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support