Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The specific function of NXNL2 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nucleoredoxin-like protein 2; RdCVF2; Rod-derived cone viability factor 2
Gene Aliases: C9orf121; NXNL2; RDCVF2
UniProt ID: (Human) Q5VZ03
Entrez Gene ID: (Human) 158046
Molecular Function:
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support