Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: TREEVVTPT LPEHTATRSQ MATSLDFKTY VDQACRAAEE FVNIYYETMD KRRRALTRLY LDKATLIWNG NAVSGLDALN NFFDTLPSSE FQVNMLDCQP VHEQATQSQT TVLVVTSGTV KFDGNKQHFF NQNFLLTAQS TPNNTVWKIA SDCFRFQDWS SS (37-197 aa encoded by BC014888)
Regulator of protein export for NES-containing proteins. Also plays a role in mRNA nuclear export.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NTF2-related export protein 2; nuclear transport factor 2-like export factor 2; Protein p15-2
Gene Aliases: 6330587F24Rik; AI787442; BM-025; DC9; NXT2; P15-2; RGD1560204
UniProt ID: (Human) Q9NPJ8, (Rat) B2GV77, (Mouse) Q3UNA4
Entrez Gene ID: (Human) 55916, (Rat) 315352, (Mouse) 237082
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support