Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LRVKLTVAPD GSQSKRSVLV NQADLQSARP RAETKPTEDL RSWEKKQRAP LRFQGDSRLE QSGCYHHCVD ENIE
Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dvl-binding protein; hNkd; hNkd1; naked cuticle-1; Naked-1; Protein naked cuticle homolog 1
Gene Aliases: Naked1; NKD; NKD1; PP7246
UniProt ID: (Human) Q969G9
Entrez Gene ID: (Human) 85407
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support