Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Peptide Sequence: NTKTSYHTPA DMLSVTAAKD AQANITNTNY KHLIHKYILL PDAMNIELTR NMNRIQSDNE YKQDYNEWYK GLGWSPAGSL EVEKAKKATE YASDQKYRQH PSNFQFKKLT
This gene encodes nebulin, a giant protein component of the cytoskeletal matrix that coexists with the thick and thin filaments within the sarcomeres of skeletal muscle. In most vertebrates, nebulin accounts for 3 to 4% of the total myofibrillar protein. The encoded protein contains approximately 30-amino acid long modules that can be classified into 7 types and other repeated modules. Protein isoform sizes vary from 600 to 800 kDa due to alternative splicing that is tissue-, species-, and developmental stage-specific. Of the 183 exons in the nebulin gene, at least 43 are alternatively spliced, although exons 143 and 144 are not found in the same transcript. Mutations in this gene are associated with recessive nemaline myopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686C1456; FLJ11505; FLJ36536; FLJ39568; FLJ39584; Nebulin; nemaline myopathy type 2
Gene Aliases: NEB; NEB177D; NEM2
UniProt ID: (Human) P20929
Entrez Gene ID: (Human) 4703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support