Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
FIGURE: 1 / 1
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Neuregulin-CRD refers to the cysteine-rich domain (CRD) of neuregulins, a family of signaling proteins involved in various cellular processes, including growth, differentiation, and development. The cysteine-rich domain is critical for the interaction of neuregulins with their receptors, such as the ErbB family of receptor tyrosine kinases, facilitating the activation of downstream signaling pathways. This domain plays a vital role in the proper functioning of neuregulins in the nervous system, heart, and other tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor-inducing activity; glial growth factor 2; heregulin, alpha (45kD, ERBB2 p185-activator); neu differentiation factor; neuregulin 1 type III beta 3; Pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor
Gene Aliases: ARIA; GGF; GGF2; HGL; HRG; HRG1; HRGA; MST131; MSTP131; NDF; NRG1; NRG1-IT2; SMDF
UniProt ID: (Human) Q02297
Entrez Gene ID: (Human) 3084, (Rat) 112400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support