Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Sequence of this protein is as follows: ASSLGAHHIH HFHGSSKH
Neuronal cell surface protein that may be involved in cell recognition and cell adhesion by forming intracellular junctions through binding to neuroligins. May play a role in formation or maintenance of synaptic junctions. May mediate intracellular signaling. May play a role in angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-latrotoxin receptor (calcium-dependent); DKFZp313P2036; FLJ35941; neurexin 1; Neurexin 1alpha; neurexin I; Neurexin I-alpha; Neurexin I-beta; Neurexin-1; Neurexin-1-alpha; Neurexin-1-beta; non-processed neurexin I-alpha
Gene Aliases: 1700062G21Rik; 9330127H16Rik; A230068P09Rik; Hs.22998; KIAA0578; mKIAA0578; NRXN1; Nrxn1b; PTHSL2; SCZD17
UniProt ID: (Human) Q9ULB1, (Mouse) Q9CS84, (Rat) Q63372
Entrez Gene ID: (Human) 9378, (Mouse) 18189, (Rat) 60391
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support