Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Reconstitute at 0.2 mg/mL in sterile PBS.
chondroitin sulfate proteoglycan that may play a role in cell adhesion and cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 245 kDa early postnatal core glycoprotein; Chondroitin sulfate proteoglycan 3; chondroitin sulfate proteoglycan 3, related sequence; Neurocan core protein; TGFbeta-induced transcript, brain
Gene Aliases: C230035B04; Cspg3; Cspg3-rs; Ncan; Tgfbit
UniProt ID: (Mouse) P55066
Entrez Gene ID: (Rat) 58982, (Mouse) 13004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support