Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5'-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 7 transmembrane receptor; neurokinin 2 receptor; Neurokinin A receptor; NK-2 receptor; NK-2R; NK2; seven transmembrane helix receptor; SK receptor; SKR; substance K receptor; Substance-K receptor; Tachykinin 2 (substance K) receptor; tachykinin NK-2 receptor; Tachykinin receptor 2
Gene Aliases: NK2R; NKNAR; SKR; TAC2R; TACR2
UniProt ID: (Human) P21452, (Mouse) P30549, (Rat) P16610
Entrez Gene ID: (Human) 6865, (Mouse) 21337, (Rat) 25007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support