Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Immunogen sequence: KMNGTEMNLEPGSRH
Reconstitution: 25 uL, 50 uL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
May be active in signaling pathway in an autocrine or paracrine fashion in several tissues. Receptor for neuropeptide S, it may mediate its action, such as inhibitory effects, on cell growth. Involved in pathogenesis of asthma and other IgE-mediated diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 154; G protein-coupled receptor PGR14; G-protein coupled receptor 154; G-protein coupled receptor for asthma susceptibility; G-protein coupled receptor PGR14; Neuropeptide S receptor; vasopressin receptor-related receptor 1; vasotocin receptor
Gene Aliases: 9330128H10Rik; ASRT2; GPR154; GPRA; MVTR; NPSR; NPSR1; PGR14; RGD1564154; VRR1
UniProt ID: (Human) Q6W5P4, (Mouse) Q8BZP8
Entrez Gene ID: (Human) 387129, (Mouse) 319239, (Rat) 300458
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support