Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total NKX22.
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; Homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; Nkx2.2
Gene Aliases: Nkx-2.2; NKX2-2; NKX2.2; NKX2B; tinman
UniProt ID: (Human) O95096, (Mouse) P42586
Entrez Gene ID: (Human) 4821, (Rat) 366214, (Mouse) 18088
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support