Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLSPVTSTPF SVKDILRLER ERSCPAASPH PRVRKSPENF QYLRMDAEPR GSEVHNAGGG GGDRKLDGSE
Antigen sequence identity: Mouse 32%; Rat 32%
Members of the NK-2 family of homeodomain proteins are key regulators of growth and development in several tissues, including brain, heart and pancreas.Nkx-2.5, also designated cardiac specific homeobox protein (Csx), is a homolog of the Drosophila tinman protein and is essential for normal cardiovascular development. Expression of Nkx-2.5 during cardiomyogenesis is required for cardiac septation, in which a single atrium and ventricle are separated into four chambers. Nkx-2.5 binds to DNA as a monomer, a homodimer or as a heterodimer with Nkx-2.3 or Nkx-2.6, which suggests that the specific protein-protein interactions of Nkx-2.5 are involved in its transcriptional regulatory function. Nkx-2.6, also a homolog of the Drosophila tinman protein, is expressed in the caudal pharyngeal pouches,the caudal heart progenitors, the sinus venosus, the outflow tract of the heart and in a short segment of the gut between stages E8.5 and E10.5 of embryogenesis. Expression of Nkx-2.6 overlaps with that of Nkx-2.5 in the pharynx and heart. However, Nkx-2.6 mutant mice are viable and fertile, which suggests that Nkx-2.6 plays a compensatory function to Nkx-2.5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein NK-2 homolog F; homeobox protein NK2 homolog F; Homeobox protein Nkx-2.6; homeobox protein NKX2.6; NK2 transcription factor related, locus 6; tinman paralog
Gene Aliases: CSX2; CTHM; NKX2-6; NKX2F; NKX4-2
UniProt ID: (Human) A6NCS4
Entrez Gene ID: (Human) 137814
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support