Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This target displays homology in the following species: Mouse: 100%; Rat: 83%
Members of the NK-2 family of homeodomain proteins are key regulators of growth and development in several tissues, including brain, heart and pancreas.Nkx-2.5, also designated cardiac specific homeobox protein (Csx), is a homolog of the Drosophila tinman protein and is essential for normal cardiovascular development. Expression of Nkx-2.5 during cardiomyogenesis is required for cardiac septation, in which a single atrium and ventricle are separated into four chambers. Nkx-2.5 binds to DNA as a monomer, a homodimer or as a heterodimer with Nkx-2.3 or Nkx-2.6, which suggests that the specific protein-protein interactions of Nkx-2.5 are involved in its transcriptional regulatory function. Nkx-2.6, also a homolog of the Drosophila tinman protein, is expressed in the caudal pharyngeal pouches,the caudal heart progenitors, the sinus venosus, the outflow tract of the heart and in a short segment of the gut between stages E8.5 and E10.5 of embryogenesis. Expression of Nkx-2.6 overlaps with that of Nkx-2.5 in the pharynx and heart. However, Nkx-2.6 mutant mice are viable and fertile, which suggests that Nkx-2.6 plays a compensatory function to Nkx-2.5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Drosophila NK2 transcription factor related, locus 6; Homeobox protein NK-2 homolog F; Homeobox protein Nkx-2.6; NK2 transcription factor related, locus 6
Gene Aliases: AV044928; Nkx-2.6; Nkx2-6; Nkx2.6; Nkx2f; tinman; Tix
UniProt ID: (Mouse) P43688
Entrez Gene ID: (Mouse) 18092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support