Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A murine NKX2-8 was isolated from the Hepal-6 cell line and showed oligonucleotide binding competitive with fetoprotein transcription factor. Nkx2.8 bound to the active AFP promoter, and antisense inhibition of Nkx2.8 mRNA translation selectively reduced expression of both the endogenous human AFP gene and transfected reporters containing the rat AFP promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Drosophila NK2 transcription factor related, locus 9; Homeobox protein NK-2 homolog H; Homeobox protein Nkx-2.8; Homeobox protein Nkx-2.9; NK-2 homolog 8; NK-2 homolog H; NK2 transcription factor related, locus 8; NK2 transcription factor related, locus 9
Gene Aliases: NKX-2.8; Nkx-2.9; NKX2-8; Nkx2-9; NKX2.8; Nkx2.9; NKX2G; NKX2H; tinman
UniProt ID: (Human) O15522, (Mouse) O70584
Entrez Gene ID: (Human) 26257, (Rat) 299061, (Mouse) 18094
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support