Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitute with water to the desired concentration.
Lyophilized antibodies are stable for 1 year from the date of purchase if stored between -20 to -80°C. Upon reconstitution, we recommend that the solution be stored at 4°C (short- term) or at -20 to -80°Cfor long term. Repeated freeze-thaw cycles should be avoided with reconstituted products.
Aliquoting is unnecessary for -20°C storage.
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Noggin; Noggin precursor; symphalangism 1 (proximal)
Gene Aliases: NOG; SYM1; SYNS1; SYNS1A
UniProt ID: (Human) Q13253
Entrez Gene ID: (Human) 9241
Molecular Function: intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support