Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Immunogen sequence: SLYYLFSSFT LNLPWTDCGH TWNSPNCTDP KLLNGSVLGN HTKYSKYKFT PAAEFYERGV LHLHESSGIH DI
Binds to an epitope located within the peptide sequence HTKYSKYKFTPAAEF as determined by overlapping synthetic peptides.
Norepinephrine Transporter [NET, or noradrenaline transporter (NAT)] is a monoamine transporter that transports the neurotransmitter noradrenaline from the synapse back to its vesicles for storage until later use. It also appears to transport the neurotransmitter dopamine in the same way, but to a lesser degree. Studies have shown a decrease in NET levels in the locus coeruleus in patients diagnosed with major depression. Cocaine, amphetamines and many therapeutic antidepressants, such as the SNRIs (serotonin-norepinephrine reuptake inhibitors) and the tricyclic antidepressants (TCAs) act to raise noradrenaline. Furthermore, deficits in the NET gene have been associated with ADHD.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NaCl-dependent norepinephrine transporter; NE transporter; NET; neurotransmitter transporter; neurotransmitter transporter, noradrenal; noradrenaline; Norepinephrine transporter; Sodium-dependent noradrenaline transporter; solute carrier family 6 (neurotransmitter transporter), member 2; solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2; Solute carrier family 6 member 2; solute carrier family 6 member 5; transmembrane region 4..26 transmembrane region 55..77 transmembrane region 105..123 transmembrane region 136..157
Gene Aliases: NAT1; NE-T; NET; NET1; SLC6A2; SLC6A5
UniProt ID: (Human) P23975, (Mouse) O55192
Entrez Gene ID: (Human) 6530, (Rat) 83511, (Mouse) 20538
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support