Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
For maximum recovery of product, centrifuge the vial prior to removing the cap.
NXF is a member of the basic helix-loop-helix-PER (MIM 602260)-ARNT (MIM 126110)-SIM (MIM 600892) (bHLH-PAS) class of transcriptional regulators, which are involved in a wide range of physiologic and developmental events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLH PAS type transcription factor NXF; bHLH-PAS type transcription factor NXF; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; Neuronal PAS domain-containing protein 4; Neuronal PAS4; PASD10 and LE-PAS
Gene Aliases: LE-PAS; Npas4; Nxf
UniProt ID: (Rat) Q8CJH6, (Mouse) Q8BGD7
Entrez Gene ID: (Rat) 266734, (Mouse) 225872
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support