Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
FIGURE: 1 / 5
NOC4L plays a specific role in biogenesis of 40 S subunit of ribosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NOC4 protein homolog; NOC4-like protein; Nucleolar complex protein 4 homolog; Nucleolar complex-associated protein 4-like protein
Gene Aliases: NET49; NOC4; NOC4L; UTP19
UniProt ID: (Human) Q9BVI4
Entrez Gene ID: (Human) 79050
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support