Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
SDS3 is a subunit of the histone deacetylase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 62 kDa nucleoporin; DKFZp547L134; FLJ20822; FLJ43869; MGC841; nuclear pore complex 1; nuclear pore glycoprotein 62; Nuclear pore glycoprotein p62; nucleoporin 62kD; nucleoporin 62kDa; Nucleoporin Nup62; nucleoporin p62
Gene Aliases: AA589433; AI426861; AI790512; AU045898; D7Ertd649e; IBSN; Np62; NUP62; Nupc1; p62; SNDI
UniProt ID: (Human) P37198, (Rat) P17955, (Mouse) Q63850
Entrez Gene ID: (Human) 23636, (Rat) 65274, (Mouse) 18226
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support