Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MENGYTYED YKNTAEWLLS HTKHRPQVAI ICGSGLGGLT DKLTQAQIFD YGEIPNFPRS TVPGHAGRLV FGFLNGRACV MMQGRFHMYE GYPLWKVTFP VRVFHLLGVD TLVVTNAAGG LNPKFEVGDI MLIRDHINLP GFSGQNPLRG PNDERFGDRF PAMSDAYDRT MRQRALSTWK QMGEQRELQE GTYVMVAGPS FETVAECRVL QKLGADAVGM STVPEVIVAR HCGLRVFGFS LITNKVIMDY ESLEKANHEE VLAAGKQAAQ KLEQFVSILM ASIPLPDKAS (1-289 aa encoded by BC106074)
This gene is a member of the DEAH box polypeptide family. The encoded protein contains the DEAH (Asp-Glu-Ala-His) motif which is characteristic of all DEAH box proteins, and is thought to function as an ATP-dependent RNA helicase that regulates the release of spliced mRNAs from spliceosomes prior to their export from the nucleus. This protein may be required for the replication of human immunodeficiency virus type 1 (HIV-1). Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 156an; HEL-S-156an; Inosine phosphorylase; inosine phosphorylase {ECO:0000250|UniProtKB:P00491}; Inosine-guanosine phosphorylase; nucleoside phosphorylase; PNP; PNP {ECO:0000250|UniProtKB:P00491}; Purine nucleoside phosphorylase; purine nucleoside phosphorylase {ECO:0000250|UniProtKB:P00491}; purine-nucleoside phosphorylase 1; purine-nucleoside:orthophosphate ribosyltransferase
Gene Aliases: AL024301; AU015798; NP; Np-1; Np-2; PNP; Pnp1; PRO1837; PUNP
UniProt ID: (Human) P00491, (Mouse) P23492
Entrez Gene ID: (Human) 4860, (Mouse) 18950, (Rat) 290029
Molecular Function: kinase nucleotide kinase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support