Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: OATP2 and OATP8, human organic anion transporters (90 kD, 120 kD, after SDS-PAGE).
The OATPs (organic anion transporter polypeptides) are a family of transport proteins that have been found to mediate the uptake of organic ions into hepatocyte cells. These proteins fall into the solute carrier family 21A (SLC21A), and are expressed in liver, kidney, and brain. These polypeptides are involved in the excretion of potentially toxic compounds and therefore play a role in the overall detoxification system of the body. Unlike other members of the OATP family, OATP2 and 8 have been detected exclusively in the liver, and there is an 80% amino acid sequence homology between these polypeptides. Other names for OATP2 include SLC21A6, OATP-C, and LST-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Liver-specific organic anion transporter 1; Liver-specific organic anion transporter 2; liver-specific organic anion transporter 3TM13; LST-1; LST-2; OATP-2; OATP-8; OATP-C; OATP1B3; Organic anion transporter 8; organic anion transporter LST-3c; Organic anion transporter SLC21A6; Organic anion-transporting polypeptide 8; SLCO1B1; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; solute carrier family 21 (organic anion transporter), member 8; Solute carrier family 21 member 6; Solute carrier family 21 member 8; Solute carrier organic anion transporter family member 1B1; Solute carrier organic anion transporter family member 1B3; solute carrier organic anion transporter family, member 1B1; solute carrier organic anion transporter family, member 1B3
Gene Aliases: HBLRR; LST-1; LST-2; LST-3TM13; LST1; LST2; LST3; OATP-8; OATP-C; OATP1B1; OATP1B3; OATP2; OATP8; OATPC; SLC21A6; SLC21A8; SLCO1B1; SLCO1B3
UniProt ID: (Human) Q9Y6L6, (Human) Q9NPD5
Entrez Gene ID: (Human) 10599, (Human) 28234
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support