Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
OAZ1 catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. This protein play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein expression is auto-regulated by polyamine-enhanced translational frameshifting.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antizyme 1; AZ1; ODC-Az; Ornithine decarboxylase antizyme 1
Gene Aliases: AZI; OAZ; OAZ1
UniProt ID: (Human) P54368
Entrez Gene ID: (Human) 4946
Molecular Function: protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support