Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
OAZ1 catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. This protein play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein expression is auto-regulated by polyamine-enhanced translational frameshifting.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antizyme 1; AZ1; ODC-Az; Ornithine decarboxylase antizyme 1
Gene Aliases: Antizyme; AZ-1; AZ1; AZI; OAZ; OAZ1; ODC-Az; ODCAC
UniProt ID: (Human) P54368, (Mouse) P54369, (Rat) P54370
Entrez Gene ID: (Human) 4946, (Mouse) 18245, (Rat) 25502
Molecular Function: protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support