Save up to 49% with protein expression workflow promo

Save now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • OCRL Antibodies

            Abnova

            OCRL Polyclonal Antibody, MaxPab™

            View all (14) OCRL antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite OCRL Polyclonal Antibody, MaxPab™

            • Antibody Testing Data (1)
            Application
            OCRL Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            OCRL Antibody (H00004952-B01P) in WB

            Western Blot analysis of OCRL expression in transfected 293T cell line by OCRL MaxPab polyclonal antibody. Lane 1: OCRL transfected lysate(98.23 KDa). Lane 2: Non-transfected lysate. .
            View Product

            OCRL Antibody in Western Blot (WB)

            Product Details

            H00004952-B01P

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            OCRL (AAH94726.1, 1 a.a. approximately 893 a.a) full-length human protein.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.4 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Product may be used with Western Blot (Transfected lysate).

            The Isotype of this product is composed of an IgG Mixture.

            Immunogen sequence: MEPPLPVGAQ PLATVEGMEM KGPLREPCAL TLAQRNGQYE LIIQLHEKEQ HVQDIIPINS HFRCVQEAEE TLLIDIASNS GCKIRVQGDW IRERRFEIPD EEHCLKFLSA VLAAQKAQSQ LLVPEQKDSS SWYQKLDTKD KPSVFSGLLG FEDNFSSMNL DKKINSQNQP TGIHREPPPP PFSVNKMLPR EKEASNKEQP KVTNTMRKLF VPNTQSGQRE GLIKHILAKR EKEYVNIQTF RFFVGTWNVN GQSPDSGLEP WLNCDPNPPD IYCIGFQELD LSTEAFFYFE SVKEQEWSMA VERGLHSKAK YKKVQLVRLV GMMLLIFARK DQCRYIRDIA TETVGTGIMG KMGNKGGVAV RFVFHNTTFC IVNSHLAAHV EDFERRNQDY KDICARMSFV VPNQTLPQLN IMKHEVVIWL GDLNYRLGMP DANEVKSLIN KKDLQRLLKF DQLNIQRTQK KAFVDFNEGE IKFIPTYKYD SKTDRWDSSG KCRVPAWCDR ILWRGTNVNQ LNYRSHMELK TSDHKPVSAL FHIGVKVVDE RRYRKVFEDS VRIMDRMEND FLPSLELSRR EFVFENVKFR QLQKEKFQIS NNGQVPCHFS FIPKLNDSQY CKPWLRAEPF EGYLEPNETV DISLDVYVSK DSVTILNSGE DKIEDILVLH LDRGKDYFLT ISGNYLPSCF GTSLEALCRM KRPIREVPVT KLIDLEKSLL QMVPLDEGAS ERPLQVPKEI WLLVDHLFKY ACHQEDLFQT PGMQEELQQI IDCLDTSIPE TIPGSNHSVA EALLIFLEAL PEPVICYELY QRCLDSAYDP RICRQVISQL PRCHRNVFRY LMAFLRELLK FSEYNSVNAN MIATLFTSLL LRPPPNLMAR QTPSDRQRAI QFLLGFLLGS EED

            Target Information

            The inositol polyphosphate 5-phosphatases selectively remove the phosphate from the 5-position of various phosphatidylinositols, which generate second messengers in response to extracellular signals. OCRL1 is a type II 5-phosphatase that is mutated in the oculocerebrorenal syndrome of Lowe (OCRL). OCRL is a rare X-linked disorder that is characterized in part by congenital cataracts, mental retardation, muscular hypotonia and renal tubular dysfunction. OCRL1 has a high affinity for phosphatidylinositol 4,5-bisphosphate as well as inositol 1,4,5-trisphosphate and inositol 1,3,4,5-tetrakisphosphate as substrates. OCRL1 is localized to the Golgi complex and is thought to be part of the trans-Golgi network (TGN), which suggests that OCRL1 plays a role in protein sorting and trafficking within the cell.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Inositol polyphosphate 5-phosphatase OCRL; Inositol polyphosphate 5-phosphatase OCRL-1; Lowe oculocerebrorenal syndrome protein; OCRL-1; oculocerebrorenal syndrome of Lowe; Phosphatidylinositol 3,4,5-triphosphate 5-phosphatase; phosphatidylinositol polyphosphate 5-phosphatase

            View more View less

            Gene Aliases: INPP5F; LOCR; NPHL2; OCRL; OCRL-1; OCRL1

            View more View less

            UniProt ID: (Human) Q01968

            View more View less

            Entrez Gene ID: (Human) 4952

            View more View less

            Function(s)

            phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity GTPase activator activity protein binding Rac GTPase binding inositol-1,4,5-trisphosphate 5-phosphatase activity inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity inositol phosphate phosphatase activity phosphatase

            Process(es)

            in utero embryonic development lipid metabolic process phosphatidylinositol biosynthetic process signal transduction cilium assembly regulation of GTPase activity positive regulation of GTPase activity inositol phosphate metabolic process phosphatidylinositol dephosphorylation regulation of small GTPase mediated signal transduction
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-vq4gn:80/100.66.79.188:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0