Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Zeta
This product is diluted and in a ready-to-use formulation.
A recommended positive control tissue for this product is Tonsil or lymph node, however positive controls are not limited to this tissue type.
OCT-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octamer sites regulating B cell specific genes. OCT-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. OCT-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeobox protein; Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; Oct-2; Octamer-binding protein 2; Octamer-binding transcription factor 2; OTF-2; POU domain, class 2, transcription factor 2
Gene Aliases: Oct-2; OCT2; OTF2; POU2F2
UniProt ID: (Human) P09086
Entrez Gene ID: (Human) 5452
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support