Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The teneurin transmembrane protein 1 (TENM1) is a member of a family of four neuronal cell surface proteins homologous to the Drosophila pair-rule gene Ten-m. TENM1 is expressed primarily in the developing central nervous system and may be proteolytically cleaved with the intracellular domain translocating to the nucleus. TENM1 is a direct target of the homeobox transcription factor EMX2, a transcription factor thought to be important for area specification in the developing cortex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: odz (odd Oz/ten-m, Drosophila) homolog 3; odz, odd Oz/ten-m homolog 1; odz, odd Oz/ten-m homolog 1(Drosophila); Protein Odd Oz/ten-m homolog 1; Ten-1; Ten-m1; tenascin M1; Tenascin-M1; teneurin 1; Teneurin transmembrane protein 1; Teneurin-1
Gene Aliases: ODZ1; Odz2; ODZ3; RGD1561678; TCAP-1; ten-1; TEN-M1; TENM1; TNM; TNM1
UniProt ID: (Human) Q9UKZ4, (Mouse) Q9WTS4
Entrez Gene ID: (Human) 10178, (Rat) 298168, (Mouse) 23963
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support