Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Peptide sequence: VFGWRSRSSG PPATFPSSKG GGGSSYMEEM YFAWLENPQS VHKSWDSFFR
Sequence homology: Cow: 85%; Dog: 100%; Horse: 92%; Human: 100%; Pig: 100%; Rat: 85%
The function of OGDHL remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-oxoglutarate dehydrogenase complex component E1-like; 2-oxoglutarate dehydrogenase-like, mitochondrial; Alpha-ketoglutarate dehydrogenase-like; OGDC-E1-like
Gene Aliases: KIAA1290; OGDHL
UniProt ID: (Human) Q9ULD0
Entrez Gene ID: (Human) 55753
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support