Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MSSSRKDHLG ASSSEPLPVI IVGNGPSGIC LSYLLSGYTP YTKPDAIHPH PLLQRKLTEA PGVSILDQDL DYLSEGLEGR SQSPVALLFD ALLRPDTDFG GNMKSVLTWK HRKEHAIPHV VLGRNLPGGA WHSIEGSMVI LSQGQWMGLP DLEVKDWMQK KRRGLRNSRA TAGDIAHYYR DYVVKKGLGH NFVSGAVVTA VEWGTPDPSS CGAQDSSPLF QVSGFLTRNQ AQQPFSLWAR NVVLATGTFD SPARLGIPGE ALPFIHHELS ALEAATRVGA VTPASDPVLI IGAGLSAADA VLYARHYNIP VIHAFRRAVD DPGLVFNQLP KMLYPEYHKV HQMMREQSIL SPSPYEGYRS LPRHQLLCFK EDCQAVFQDL EGVEKVFGVS LVLVLIGSHP DLSFLPGAGA DFAVDPDQPL SAKRNPIDVD PFTYQSTRQE GLYAMGPLAG DNFVRFVQGG ALAVASSLLR KETRKPP
OSGIN1 regulates the differentiation and proliferation of normal cells through the regulation of cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMSC-derived growth inhibitor; Bone marrow stromal cell-derived growth inhibitor; huOKL38; Ovary, kidney and liver protein 38; Oxidative stress-induced growth inhibitor 1; Pregnancy-induced growth inhibitor OKL38
Gene Aliases: BDGI; OKL38; OSGIN1
UniProt ID: (Human) Q9BZ70
Entrez Gene ID: (Human) 29948
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support