Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NLSAVLTGI QEEIGAYDYE ELHQRVLSLE TRLRDCMKKL TCGKLMKITG PVTVKTSGTR FGAWMTDPLA SEKNNRVWYM DSYTNNKIVR EYKSIADFVS GAESRTYNLP FKWAGTNHVV YNGSLYFNKY QSNIIIKYSF DMGRVLAQRS LEYAGFHNVY PYTWGGFSDI DLMADEIGLW AVYATNQNAG NIVISQLNQD TLEVMKSWST GYPKRSAGES FMICGTLYVT NSHLTGAKVY YSYSTKTSTY EYTDIPFHNQ YFHISMLDYN ARDRALCAWN NGHQVLFNVT LFHIIKTEDD T (179-478 aa encoded by BC022531)
OLFM3 (olfactomedin 3), also known as NOE3, is a 478 amino acid protein that interacts with myocilin. Myocilin is an extracellular protein that plays a key role in the actomyosin system and is responsible for controlling intraocular pressure. OLFM3 is a secreted protein that contains an olfactomedin-like (OLF) domain, an approximately 260 amino acid motif commonly found in secreted glycoproteins. OLFM3 localizes to the Golgi apparatus of the cell and is highly expressed in both eye and brain tissue. Mutations in the gene that encodes OLFM3 may cause severe glaucoma, a condition in which increased intraocular pressure within the eyeball causes loss of eye sight.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Noelin-3; olfactomedin related ER localized protein 3; Olfactomedin-3; Optimedin
Gene Aliases: B230206G02Rik; NOE3; NOELIN3; OLFM3; OPTIMEDIN; UNQ1924/PRO4399
UniProt ID: (Human) Q96PB7, (Mouse) P63056
Entrez Gene ID: (Human) 118427, (Mouse) 229759
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support