Save up to 49% with protein expression workflow promo

Save now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • OLIG1/OLIG2/OLIG3 Antibodies

            Invitrogen

            OLIG1/OLIG2/OLIG3 Monoclonal Antibody (257224)

            1 Reference
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # MA5-23805
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            487.00
            100 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite OLIG1/OLIG2/OLIG3 Monoclonal Antibody (257224)

            Product Details

            MA5-23805

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (Frozen) (IHC (F))

            8-25 µg/mL
            -

            Immunocytochemistry (ICC/IF)

            8-25 µg/mL
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human

            Host/Isotype

            Mouse / IgG2a

            Class

            Monoclonal

            Type

            Antibody

            Clone

            257224

            Immunogen

            E. coli-derived recombinant human Olig1 (aa 1-272)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Concentration

            0.5 mg/mL

            Purification

            Protein A/G

            Storage buffer

            PBS with 5% trehalose

            Contains

            No Preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2607923

            Product Specific Information

            In direct ELISAs, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) Olig2 and 50-100% cross-reactivity with rhOlig3.

            Reconstitute at 0.5 mg/mL in sterile PBS.

            Target Information

            Olig1 promotes formation and maturation of oligodendrocytes, especially within the brain. Cooperates with OLIG2 to establish the pMN domain of the embryonic neural tube. Olig2 is an essential regulator of ventral neuroectodermal progenitor cell fate. The gene is involved in a chromosomal translocation t(14;21)(q11.2;q22) associated with T-cell acute lymphoblastic leukemia. Its chromosomal location is within a region of chromosome 21 which has been suggested to play a role in learning deficits associated with Down syndrome. Tissue specificity: Expressed in the brain, in oligodendrocytes. Strongly expressed in oligodendrogliomas, while expression is weak to moderate in astrocytomas. Expression in glioblastomas highly variable.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: basic domain, helix-loop-helix protein, class B, 1; basic domain, helix-loop-helix protein, class B, 6; bHLHb1; bHLHb6; bHLHb7; bHLHe19; bHLHe20; bHLHe21; Class B basic helix-loop-helix protein 1; Class B basic helix-loop-helix protein 6; Class B basic helix-loop-helix protein 7; Class E basic helix-loop-helix protein 19; Class E basic helix-loop-helix protein 20; Class E basic helix-loop-helix protein 21; human protein kinase C-binding protein RACK17; olig-2; Oligo1; Oligo2; Oligo2 antibody; Oligo3; oligodendrocyte lineage transcription factor 1; Oligodendrocyte transcription factor 1; Oligodendrocyte transcription factor 2; Oligodendrocyte transcription factor 3; oligodendrocyte-specific bHLH transcription factor 1; oligodendrocyte-specific bHLH transcription factor 2; Protein kinase C-binding protein 2; Protein kinase C-binding protein RACK17

            View more View less

            Gene Aliases: BHLHB1; BHLHB6; BHLHB7; BHLHE19; BHLHE20; BHLHE21; OLIG1; OLIG2; OLIG3; OLIGO2; PRKCBP2; RACK17

            View more View less

            UniProt ID: (Human) Q8TAK6, (Human) Q13516, (Human) Q7RTU3

            View more View less

            Entrez Gene ID: (Human) 116448, (Human) 10215, (Human) 167826

            View more View less

            Function(s)

            DNA binding protein binding protein dimerization activity transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding protein homodimerization activity HMG box domain binding RNA polymerase II transcription corepressor activity basic helix-loop-helix transcription factor

            Process(es)

            transcription, DNA-templated regulation of transcription, DNA-templated neuron fate commitment oligodendrocyte differentiation negative regulation of transcription from RNA polymerase II promoter spinal cord motor neuron differentiation spinal cord oligodendrocyte cell fate specification thalamus development myelination negative regulation of neuron differentiation positive regulation of oligodendrocyte differentiation spinal cord motor neuron cell fate specification spinal cord motor neuron migration
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-4g5vm:80/100.66.76.159:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0