Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAQQWSLQRL AGRHPQDSYE DSTQSSIFTY TNSNSTRGPF EGPNYHIAPR WV
Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal. May increase spectral sensitivity in dim light. Expresssed in cone photoreceptor cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cone dystrophy 5 (X-linked); GOP; Green cone photoreceptor pigment; green cone pigment; green long wavelength sensitive cone opsin; green LWS photopigment; Green-sensitive opsin; M opsin; Medium wavelength-sensitive cone opsin; Medium-wave-sensitive opsin 1; Medium-wave-sensitive opsin 2; Medium-wave-sensitive opsin 3; midwavelength sensitive opsin; MWS opsin; opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan), green opsin; Opsin 1 cone pigments medium-wave-sensitive 2; opsin 1 cone pigments medium-wave-sensitive 3; photopigment apoprotein
Gene Aliases: CBBM; CBD; COD5; GCP; GOP; OPN1MW; OPN1MW1; OPN1MW2; OPN1MW3
UniProt ID: (Human) P0DN77, (Mouse) O35599, (Rat) O35476
Entrez Gene ID: (Human) 2652, (Mouse) 14539, (Rat) 89810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support