Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ994; Hs6M1-16; Olfactory receptor 2H1; Olfactory receptor 2H6; Olfactory receptor 2H8; Olfactory receptor 6-2; Olfactory receptor OR6-32; olfactory receptor, family 2, subfamily H, member 1; olfactory receptor, family 2, subfamily H, member 6; olfactory receptor, family 2, subfamily H, member 8; OLFR42A-9004.14/9026.2; OR6-2
Gene Aliases: 6M1-16; dJ994E9.4; HS6M1-16; OLFR42A-9004-14; OLFR42A-9004.14/9026.2; OR2H1; OR2H6; OR2H8; OR6-2
UniProt ID: (Human) Q9GZK4
Entrez Gene ID: (Human) 26716
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support