Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: novel 7 transmembrane receptor (rhodopsin family) protein; Olfactory receptor 2M3; Olfactory receptor 2M6; Olfactory receptor OR1-54; olfactory receptor, family 2, subfamily M, member 3 pseudogene; olfactory receptor, family 2, subfamily M, member 6
Gene Aliases: OR1-54; OR2M3; OR2M3P; OR2M6; OST003
UniProt ID: (Human) Q8NG83
Entrez Gene ID: (Human) 127062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support