Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Monkey (94%); Marmoset, Mouse, Hamster, Rabbit (88%); Gibbon, Galago, Rat (82%).
IHC-P with human FFPE tissues requires proteinase K digestion.
Odorant receptor (Potential).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-GPCR; Dresden-G-protein-coupled receptor; G-protein coupled receptor 164; Olfactory receptor 51E1; Olfactory receptor 52A3; olfactory receptor OR11-15; olfactory receptor, family 51, subfamily E, member 1 pseudogene; olfactory receptor, family 52, subfamily A, member 3 pseudogene; prostate overexpressed G protein-coupled receptor; Prostate-overexpressed G protein-coupled receptor; Prostate-specific G protein-coupled receptor 2
Gene Aliases: D-GPCR; DGPCR; GPR136; GPR164; OR51E1; OR51E1P; OR52A3P; POGR; PSGR2
UniProt ID: (Human) Q8TCB6
Entrez Gene ID: (Human) 143503
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support