Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Hamster, Elephant, Rabbit, Opossum (100%) Marmoset, Dog (93%).
Olfactory receptors function by associating with odorant molecules to launch a neuronal response that elicits the perception of smell. Olfactory receptor genes are unevenly dispersed among 51 loci on 21 human chromosomes, and are composed of 172 subfamilies. OR6N1 (olfactory receptor, family 6, subfamily N, member 1), also known as olfactory receptor OR1-22, is a 312 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 1 family. As the largest family in the genome, the G-protein coupled receptor 1 family originated from single coding-exon genes. Encoded by a gene that maps to human chromosome 1q23.1, OR6N1 functions as an odorant receptor. Like many hormone and neurotransmitter receptors, OR6N1 shares a 7-transmembrane domain structure and recognizes G protein-mediated transduction of odorant signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Olfactory receptor 6N1; olfactory receptor OR1-22; olfactory receptor, family 6, subfamily N, member 1
Gene Aliases: OR1-22; OR6N1
UniProt ID: (Human) Q8NGY5
Entrez Gene ID: (Human) 128372
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support