Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Despite the predicted molecular weight, ORAI1 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human ovary tissue lysate.
PA5-20322 can be used with blocking peptide PEP-0442.
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium release-activated calcium channel 1; Calcium release-activated calcium channel protein 1; calcium release-activated calcium modulator 1; FLJ14466; Protein orai-1; Transmembrane protein 142A
Gene Aliases: CRACM1; D730049H07Rik; IMD9; orai-1; ORAI1; ORAT1; RGD1311873; TAM2; TMEM142A
UniProt ID: (Human) Q96D31, (Rat) Q5M848, (Mouse) Q8BWG9
Entrez Gene ID: (Human) 84876, (Rat) 304496, (Mouse) 109305
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support