Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with bovine, rat and Xenopus and zebrafish based on sequence homology.
Negative regulator of sphingolipid synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adoplin-2; expressed in normal aorta; ORM1-like protein 2
Gene Aliases: adoplin-2; HSPC160; MST095; MSTP095; ORMDL2
UniProt ID: (Human) Q53FV1
Entrez Gene ID: (Human) 29095
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support